N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester
Supplier: Apollo Scientific
OR346047-1G OR346047-500MG
APOSOR346047-1GEA 0 EUR
APOSOR346047-1G APOSOR346047-500MG
N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester
N'-[1-(2-Methoxyethylamino)-2-phenoxyethylidene]hydrazine carboxylic acid-tert-butyl ester
Formula: C₁₆H₂₅N₃O₄
MW: 323.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568262
CAS Number: 1053658-05-1