Methyl-5-hydroxymethyl-2-methyl-2H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-(hydroxymethyl)-1-methyl-1H-pyrazole-5-carboxylate
OR346080-1G OR346080-5G
APOSOR346080-1GEA 243 EUR
APOSOR346080-1G APOSOR346080-5G
Methyl-5-hydroxymethyl-2-methyl-2H-pyrazole-3-carboxylate
Methyl-5-hydroxymethyl-2-methyl-2H-pyrazole-3-carboxylate
Formula: C₇H₁₀N₂O₃
MW: 170.17 g/mol
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD10568327
CAS Number: 1208081-25-7