N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine
Supplier: Apollo Scientific
OR346112-1G OR346112-5G
APOSOR346112-1GEA 0 EUR
APOSOR346112-1G APOSOR346112-5G
N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine
N-Hydroxy-2-morpholin-4-yl-2-phenylacetamidine
Formula: C₁₂H₁₇N₃O₂
MW: 235.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052466
CAS Number: 1053659-35-0