2-Amino-5-chloropyridine-3-sulphonyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR346130-1G
OR346130-250MG
OR346130-5G
APOSOR346130-1GEA
227
EUR
APOSOR346130-1G
APOSOR346130-250MG
APOSOR346130-5G
2-Amino-5-chloropyridine-3-sulphonyl chloride
2-Amino-5-chloropyridine-3-sulphonyl chloride
Formula:
C₅H₄Cl₂N₂O₂S MW: 227.07 g/mol Melting Pt: 117…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD12402514 CAS Number: 163137-45-9 UN: 3261 ADR: 8 II |