5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester
Supplier: Apollo Scientific
Warning
OR346170-1G OR346170-500MG
APOSOR346170-1GEA 0 EUR
APOSOR346170-1G APOSOR346170-500MG
5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester
5-Chloromethyl-2-methyl-2H-pyrazole-3-carboxylic acid methyl ester
Formula: C₇H₉ClN₂O₂
MW: 188.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845756
CAS Number: 1208081-34-8