6-Phenylpyrimidine-2,4-diol
Supplier: Apollo Scientific
OR346181-1G OR346181-500MG OR346181-5G
APOSOR346181-1GEA 0 EUR
APOSOR346181-1G APOSOR346181-500MG APOSOR346181-5G
6-Phenylpyrimidine-2,4-diol
6-Phenylpyrimidine-2,4-diol
Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00128171
CAS Number: 13345-09-0