2-Chloro-6-methoxypyridine-3-sulphonyl chloride
Supplier: Apollo Scientific
Danger
OR346193-1G OR346193-500MG
APOSOR346193-1GEA 0 EUR
APOSOR346193-1G APOSOR346193-500MG
2-Chloro-6-methoxypyridine-3-sulphonyl chloride
2-Chloro-6-methoxypyridine-3-sulphonyl chloride
Formula: C₆H₅Cl₂NO₃S
MW: 242.08 g/mol
Melting Pt: 94…95 °C
Storage Temperature: Ambient
MDL Number: MFCD15142789
CAS Number: 1208081-26-8
UN: 3261
ADR: 8 II