2-Chloro-5-methylpyridine-3-sulphonyl chloride
Supplier: Apollo Scientific
OR346213-1G OR346213-500MG
APOSOR346213-1GEA 0 EUR
APOSOR346213-1G APOSOR346213-500MG
2-Chloro-5-methylpyridine-3-sulphonyl chloride
2-Chloro-5-methylpyridine-3-sulphonyl chloride
Formula: C₆H₅Cl₂NO₂S
MW: 226.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15142818
CAS Number: 1208081-98-4