1-(5-Chloro-2-pyridinyl)piperazine
Supplier: Apollo Scientific
Synonyms: 1-(5-Chloro-2-pyridyl)piperazine
OR346244-1G OR346244-5G
APOSOR346244-1GEA 0 EUR
APOSOR346244-1G APOSOR346244-5G
1-(5-Chloro-2-pyridinyl)piperazine
1-(5-Chloro-2-pyridinyl)piperazine
Formula: C₉H₁₂ClN₃
MW: 197.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115049
CAS Number: 87394-65-8