1-Bromo-3-methylbutan-2-one
Supplier: Apollo Scientific
Danger
OR346253-1G OR346253-5G
APOSOR346253-1GEA 119 EUR
APOSOR346253-1G APOSOR346253-5G
1-Bromo-3-methylbutan-2-one
1-Bromo-3-methylbutan-2-one
Formula: C₅H₉BrO
MW: 165.0284 g/mol
Boiling Pt: 74…75 °C/46mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00015716
CAS Number: 19967-55-6
UN: 3265
ADR: 8,III