2-(Chloromethyl)benzoyl chloride
Supplier: Apollo Scientific
Danger
OR346332-5G
APOSOR346332-5GEA 77 EUR
APOSOR346332-5G
2-(Chloromethyl)benzoyl chloride
2-(Chloromethyl)benzoyl chloride
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Melting Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00152348
CAS Number: 42908-86-1
EINECS: 255-998-2
UN: 2927
ADR: 6.1+8,II