Tetrahydro-2H-pyran-2-carbonyl chloride
Supplier: Apollo Scientific
Danger
OR346346-1G OR346346-250MG
APOSOR346346-1GEA 470 EUR
APOSOR346346-1G APOSOR346346-250MG
Tetrahydro-2H-pyran-2-carbonyl chloride
Tetrahydro-2H-pyran-2-carbonyl chloride
Formula: C₆H₉ClO₂
Boiling Pt: 55 °C
Storage Temperature: Ambient
CAS Number: 40053-81-4
UN: 3265
ADR: 8 II