2,6-Dibromo-3-hydroxypyridine 95+%
Supplier: Apollo Scientific
Danger
OR346545-1G OR346545-5G
APOSOR346545-1GEA 24.4 EUR
APOSOR346545-1G APOSOR346545-5G
2,6-Dibromo-3-hydroxypyridine 95+%
2,6-Dibromo-3-hydroxypyridine
Formula: C₅H₃Br₂NO
Melting Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD01318479
CAS Number: 6602-33-1
UN: 2811
ADR: 6.1,III