α,α-Dimethoxytoluene
Supplier: Apollo Scientific
Warning
Synonyms: Benzaldehyde dimethyl acetal
OR3467-100G OR3467-500G OR3467-1KG
APOSOR3467-100GEA 21.9 EUR
APOSOR3467-100G APOSOR3467-500G APOSOR3467-1KG
α,α-Dimethoxytoluene
α,α-Dimethoxytoluene
Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 196 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00008491
CAS Number: 1125-88-8
EINECS: 214-413-0