5-(Chloromethyl)-3-isopropylisoxazole 95+%

Supplier: Apollo Scientific

OR346708-1G
APOSOR346708-1GEA 318 EUR
APOSOR346708-1G
5-(Chloromethyl)-3-isopropylisoxazole 95+%
5-(Chloromethyl)-3-isopropylisoxazole
Formula: C₇H₁₀ClNO
MW: 159.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701716
CAS Number: 64988-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR