Benzo[d][1,3]dioxol-4-amine
Supplier: Apollo Scientific
Warning
OR350054-1G
APOSOR350054-1GEA 66.7 EUR
APOSOR350054-1G
Benzo[d][1,3]dioxol-4-amine
Benzo[d][1,3]dioxol-4-amine
Formula: C₇H₇NO₂
MW: 137.1400 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD09038121
CAS Number: 1668-84-4