4-Nitrobenzophenone 97%
Supplier: Apollo Scientific
Synonyms: p-Nitrobenzophenone
OR350288-100G OR350288-25G OR350288-500G
APOSOR350288-100GEA 79.9 EUR
APOSOR350288-100G APOSOR350288-25G APOSOR350288-500G
4-Nitrobenzophenone 97%
4-Nitrobenzophenone
Formula: C₁₃H₉NO₃
MW: 227.22 g/mol
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007354
CAS Number: 1144-74-7