2-Naphthaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: β-Naphthaldehyde
OR350292-100G
APOSOR350292-100GEA 114 EUR
APOSOR350292-100G
2-Naphthaldehyde
2-Naphthaldehyde
Formula: C₁₁H₈O
MW: 156.18 g/mol
Boiling Pt: 160…19mm °C
Melting Pt: 58…61 °C
Flash Pt: >100 °C
Storage Temperature: Freezer
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2
UN: 3335
ADR: 9,III