3-Nitrobenzylamine hydrochloride 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Nitrobenzylammonium hydrochloride
OR350302-25G OR350302-5G OR350302-100G
APOSOR350302-25GEA 180 EUR
APOSOR350302-25G APOSOR350302-5G APOSOR350302-100G
3-Nitrobenzylamine hydrochloride 98%
3-Nitrobenzylamine hydrochloride
Formula: C₇H₉ClN₂O₂
MW: 188.62 g/mol
Melting Pt: 226…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00012858
CAS Number: 26177-43-5
EINECS: 247-502-8