O-(Tetrahydropyran-2-yl)-hydroxylamine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Aminooxy)tetrahydro-2H-pyran, N-(2-tetrahydropyranyloxy)amine, O-(Tetrahydro-2H-pyran-2-yl)hydroxylamine
OR350311-25G OR350311-100G OR350311-5G
APOSOR350311-25GEA 159 EUR
APOSOR350311-25G APOSOR350311-100G APOSOR350311-5G
O-(Tetrahydropyran-2-yl)-hydroxylamine 98%
O-(Tetrahydropyran-2-yl)-hydroxylamine
Formula: C₅H₁₁NO₂
MW: 117.148 g/mol
Boiling Pt: 81 °C/20mmHg
Melting Pt: 34…37 °C
Flash Pt: 82 °C
Storage Temperature: Freezer
MDL Number: MFCD01321374
CAS Number: 6723-30-4