5,6,7,8-Tetrahydroquinoline 97%
Supplier: Apollo Scientific
Warning
OR350369-100G OR350369-25G OR350369-500G
APOSOR350369-100GEA 120 EUR
APOSOR350369-100G APOSOR350369-25G APOSOR350369-500G
5,6,7,8-Tetrahydroquinoline 97%
5,6,7,8-Tetrahydroquinoline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 222 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006734
CAS Number: 10500-57-9