2,3,4,5-Tetramethyl-2-cyclopentenone 95%
Supplier: Apollo Scientific
Synonyms: 2,3,4,5-Tetramethylcyclopent-2-enone
OR3509-5G OR3509-25G
APOSOR3509-5GEA 26.1 EUR
APOSOR3509-5G APOSOR3509-25G
2,3,4,5-Tetramethyl-2-cyclopentenone 95%
2,3,4,5-Tetramethyl-2-cyclopentenone
Formula: C₉H₁₄O
MW: 138.21 g/mol
Boiling Pt: 100 °C/30mmHg
Density: 0.917 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6