3-Bromo-5-chlorobenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR3540-10G OR3540-25G OR3540-5G
APOSOR3540-10GEA 24.8 EUR
APOSOR3540-10G APOSOR3540-25G APOSOR3540-5G
3-Bromo-5-chlorobenzoic acid 98%
3-Bromo-5-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 189…194 °C
Storage Temperature: Ambient
MDL Number: MFCD06797227
CAS Number: 42860-02-6