Ethyl-5-oxazolecarboxylate
Supplier: Apollo Scientific
Warning
OR3553-5G OR3553-25G
APOSOR3553-5GEA 23.2 EUR
APOSOR3553-5G APOSOR3553-25G
Ethyl-5-oxazolecarboxylate
Ethyl 5-oxazolecarboxylate
Formula: C₆H₇NO₃
Boiling Pt: 60 °C/2.25mmHg
Density: 1.163 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Freezer
MDL Number: MFCD04114930
CAS Number: 118994-89-1