2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulphonamide 95+%
Supplier: Apollo Scientific
Warning
Synonyms: N,N-Dimethylindoline-5-sulphonamide
OR3558-10G OR3558-1G OR3558-5G
APOSOR3558-10GEA 828 EUR
APOSOR3558-10G APOSOR3558-1G APOSOR3558-5G
2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulphonamide 95+%
2,3-Dihydro-N,N-dimethyl-5(1H)-indolesulphonamide
Formula: C₁₀H₁₄N₂O₂S
MW: 226.3 g/mol
Melting Pt: 108…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00214476
CAS Number: 99169-99-0