1,3-Dimethoxy-4-nitrobenzene
Supplier: Apollo Scientific
Warning
Synonyms: 2,4-Dimethoxynitrobenzene, 2,4-Dimethoxy-1-nitrobenzene
OR3561-100G OR3561-25G OR3561-5G
APOSOR3561-100GEA 174 EUR
APOSOR3561-100G APOSOR3561-25G APOSOR3561-5G
1,3-Dimethoxy-4-nitrobenzene
1,3-Dimethoxy-4-nitrobenzene
Formula: C₈H₉NO₄
MW: 183.16 g/mol
Melting Pt: 74…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00024210
CAS Number: 4920-84-7