Methyl-2-methoxy-5-(methylsulphonyl)benzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-methoxy-5-(methylsulphonyl)benzoate, 2-Methoxy-5-(methylsulphonyl)benzoic acid Methyl Ester
OR3570-100G OR3570-25G OR3570-5G
APOSOR3570-100GEA 104 EUR
APOSOR3570-100G APOSOR3570-25G APOSOR3570-5G
Methyl-2-methoxy-5-(methylsulphonyl)benzoate
Methyl-2-methoxy-5-(methylsulphonyl)benzoate
Formula: C₁₀H₁₂O₅S
MW: 244.26 g/mol
Melting Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD01317545
CAS Number: 63484-12-8