3,5-Dichlorophenylboronic acid pinacol ester 97%
Supplier: Apollo Scientific
Synonyms: 2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR360018-25G
APOSOR360018-25GEA 150 EUR
APOSOR360018-25G
3,5-Dichlorophenylboronic acid pinacol ester 97%
3,5-Dichlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BCl₂O₂
MW: 272.97 g/mol
Melting Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD05863906
CAS Number: 68716-51-8