3-(Morpholino)phenylboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
(3-Morpholinophenyl)boronic acid
OR360139-5G
APOSOR360139-5GEA
178
EUR
APOSOR360139-5G
3-(Morpholino)phenylboronic acid 97%
3-(Morpholino)phenylboronic acid
Formula:
C₁₀H₁₄BNO₃ MW: 207.0341 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03412095 CAS Number: 863377-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...