2-Isopropoxy-5-pyridineboronic acid pinacol ester 98%
Supplier: Apollo Scientific
Synonyms: 2-Isopropoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(6-Isopropoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Isopropoxy-3-pyridylboronic acid Pinacol ester, 2-Isopropoxypyridine-5-boronic acid pinacol ester
OR360292-5G
APOSOR360292-5GEA 212 EUR
APOSOR360292-5G
2-Isopropoxy-5-pyridineboronic acid pinacol ester 98%
2-Isopropoxy-5-pyridineboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368871
CAS Number: 871839-91-7