4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,1,3-Benzothiadiazole-4,7-diboronic Acid Bis(pinacol) Ester, 4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]thiadiazole
OR360668-1G OR360668-5G
APOSOR360668-1GEA 55.1 EUR
APOSOR360668-1G APOSOR360668-5G
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole 98%
4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Formula: C₁₈H₂₆B₂N₂O₄S
MW: 388.10 g/mol
Melting Pt: 207…212 °C
Storage Temperature: Ambient
MDL Number: MFCD11656086
CAS Number: 934365-16-9