Propen-1-ylboronic acid pinacol ester 98%
Supplier: Apollo Scientific
Warning
OR361552-1G OR361552-5G
APOSOR361552-1GEA 377 EUR
APOSOR361552-1G APOSOR361552-5G
Propen-1-ylboronic acid pinacol ester 98%
Propen-1-ylboronic acid pinacol ester
Formula: C₉H₁₇BO₂
Density: 0.881 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD15143598
CAS Number: 72824-05-6
UN: 1993
ADR: 3 III