4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2-(4-(2-Bromoethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR3628-5G
APOSOR3628-5GEA 217 EUR
APOSOR3628-5G
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester 96%
4-(2-Bromoethoxy)benzeneboronic acid pinacol ester
Formula: C₁₄H₂₀BBrO₃
MW: 327.03 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD09027285
CAS Number: 913836-27-8