Methyl-4-formylbenzoate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate
OR3659-100G OR3659-25G
APOSOR3659-100GEA 40.6 EUR
APOSOR3659-100G APOSOR3659-25G
Methyl-4-formylbenzoate
Methyl 4-formylbenzoate
Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 265 °C
Melting Pt: 60…63 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00006950
CAS Number: 1571-08-0
EINECS: 216-385-5