7-Bromoindole 97%
Supplier: Apollo Scientific
Warning
Synonyms: 7-Bromo-1H-indole
OR3667-5G OR3667-1G
APOSOR3667-5GEA 42.2 EUR
APOSOR3667-5G APOSOR3667-1G
7-Bromoindole 97%
7-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 41…44 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00799492
CAS Number: 51417-51-7