2-Iodopyrazine
Supplier: Apollo Scientific
Warning
OR3669-1G OR3669-25G OR3669-5G
APOSOR3669-1GEA 21.9 EUR
APOSOR3669-1G APOSOR3669-25G APOSOR3669-5G
2-Iodopyrazine
2-Iodopyrazine
Formula: C₄H₃IN₂
MW: 205.99 g/mol
Boiling Pt: 109…110 °C/34mmHg
Density: 2.086 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319019
CAS Number: 32111-21-0