7-Bromo-1-methyl-5-nitro-1H-indazole
Supplier: Apollo Scientific
Warning
OR3677-1G OR3677-500MG
APOSOR3677-1GEA 671 EUR
APOSOR3677-1G APOSOR3677-500MG
7-Bromo-1-methyl-5-nitro-1H-indazole
7-Bromo-1-methyl-5-nitro-1H-indazole
Formula: C₈H₆BrN₃O₂
MW: 256.561 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD09607926
CAS Number: 952183-39-0