1-Methyl-1H-indazol-6-amine
Supplier: Apollo Scientific
Warning
Synonyms: 6-Amino-1-methyl-1H-indazole
OR3690-500MG OR3690-1G OR3690-5G
APOSOR3690-500MGEA 31.9 EUR
APOSOR3690-500MG APOSOR3690-1G APOSOR3690-5G
1-Methyl-1H-indazol-6-amine
1-Methyl-1H-indazol-6-amine
Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 174…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00456076
CAS Number: 74728-65-7