2-(Tributylstannyl)pyridine 95%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Tri-n-butylstannyl)pyridine
OR3779-25G OR3779-100G OR3779-5G
APOSOR3779-25GEA 0 EUR
APOSOR3779-25G APOSOR3779-100G APOSOR3779-5G
2-(Tributylstannyl)pyridine 95%
2-(Tributylstannyl)pyridine
Remainder tributyltin halide.
Formula: C₁₇H₃₁NSn
MW: 368.15 g/mol
Boiling Pt: 132…134 °C/1mmHg
Density: 1.137 g/cm³ (25 °C)
Flash Pt: 24 °C
Storage Temperature: Freezer
MDL Number: MFCD00052052
CAS Number: 17997-47-6
UN: 2929
ADR: 6.1+3 II