[3-(1H-Pyrazol-1-yl)phenyl]methanol 96%
Supplier: Apollo Scientific
Warning
Synonyms: (3-(1H-Pyrazol-1-yl)phenyl)methanol, 3-(1H-Pyrazol-1-yl)benzyl alcohol
OR3816-1G
APOSOR3816-1GEA 249 EUR
APOSOR3816-1G
[3-(1H-Pyrazol-1-yl)phenyl]methanol 96%
[3-(1H-Pyrazol-1-yl)phenyl]methanol
Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD06740322
CAS Number: 864068-80-4