3-Chloropropene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Chloro-1-propene, Allyl chloride
OR3848-2.5L OR3848-500ML
APOSOR3848-2.5LEA 186 EUR
APOSOR3848-2.5L APOSOR3848-500ML
3-Chloropropene 98%
3-Chloropropene
Formula: C₃H₅Cl
MW: 76.53 g/mol
Boiling Pt: 44…46 °C
Melting Pt: –135 °C
Density: 0.939 g/cm³ (25 °C)
Flash Pt: –28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000984
CAS Number: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3+6.1,I