2,7-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Warning
OR3867-1G OR3867-5G OR3867-100MG
APOSOR3867-1GEA 250 EUR
APOSOR3867-1G APOSOR3867-5G APOSOR3867-100MG
2,7-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
2,7-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00139513
CAS Number: 81438-53-1