2-Nitrochalcone
Supplier: Apollo Scientific
Warning
OR3872-5G OR3872-25G
APOSOR3872-5GEA 114 EUR
APOSOR3872-5G APOSOR3872-25G
2-Nitrochalcone
2-Nitrochalcone
Formula: C₁₅H₁₁NO₃
MW: 253.26 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00031069
CAS Number: 7473-93-0