4-Piperidinemethanol 97%
Supplier: Apollo Scientific
Danger
Synonyms: Piperidin-4-ylmethanol
OR3906-100G OR3906-500G
APOSOR3906-100GEA 145 EUR
APOSOR3906-100G APOSOR3906-500G
4-Piperidinemethanol 97%
4-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 118…120 °C
Melting Pt: 55…59 °C
Density: 0.997 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00174228
CAS Number: 6457-49-4
UN: 3263
ADR: 8,III