3-Bromoisatoic anhydride
Supplier: Apollo Scientific
Warning
Synonyms: 8-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione
OR400020-1G OR400020-5G OR400020-25G
APOSOR400020-1GEA 34.8 EUR
APOSOR400020-1G APOSOR400020-5G APOSOR400020-25G
3-Bromoisatoic anhydride
3-Bromoisatoic anhydride
Formula: C₈H₄BrNO₃
MW: 242.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738735
CAS Number: 331646-98-1