7-Bromobenzofuran-3(2H)-one
Supplier: Apollo Scientific
OR400051-1G OR400051-250MG
APOSOR400051-1GEA 277 EUR
APOSOR400051-1G APOSOR400051-250MG
7-Bromobenzofuran-3(2H)-one
7-Bromobenzofuran-3(2H)-one
Formula: C₈H₅BrO₂
MW: 213.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03160589
CAS Number: 519018-52-1