5-Iodo-2-methoxybenzaldehyde
Supplier: Apollo Scientific
Danger
OR400057-1G OR400057-5G OR400057-25G
APOSOR400057-1GEA 21.9 EUR
APOSOR400057-1G APOSOR400057-5G APOSOR400057-25G
5-Iodo-2-methoxybenzaldehyde
5-Iodo-2-methoxybenzaldehyde
Formula: C₈H₇IO₂
MW: 262.046 g/mol
Melting Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD04128999
CAS Number: 42298-41-9
UN: 2811
ADR: 6.1 III