5'-Bromo-2'-methoxyacetophenone
Supplier: Apollo Scientific
Danger
Synonyms: 1-(5-Bromo-2-methoxyphenyl)ethanone
OR400108-25G
APOSOR400108-25GEA 134 EUR
APOSOR400108-25G
5'-Bromo-2'-methoxyacetophenone
5'-Bromo-2'-methoxyacetophenone
Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 38…39 °C
Storage Temperature: Ambient
MDL Number: MFCD03789141
CAS Number: 16740-73-1
UN: 3261
ADR: 8,III