5-Bromobenzofuran-3(2H)-one
Supplier: Apollo Scientific
|
Warning
|
OR40013-1G
OR40013-250MG
APOSOR40013-1GEA
50.9
EUR
APOSOR40013-1G
APOSOR40013-250MG
5-Bromobenzofuran-3(2H)-one
5-Bromobenzofuran-3(2H)-one
Formula:
C₈H₅BrO₂ MW: 213.0300 g/mol Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD06738724 CAS Number: 54450-20-3 |