4-Ethynylbenzonitrile

Supplier: Apollo Scientific
Warning

OR400253-1G OR400253-5G OR400253-25G
APOSOR400253-1GEA 79.9 EUR
APOSOR400253-1G APOSOR400253-5G APOSOR400253-25G
4-Ethynylbenzonitrile
4-Ethynylbenzonitrile
Formula: C₉H₅N
MW: 127.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974058
CAS Number: 3032-92-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR